Learn More
Tocris Bioscience™ Substance P
Sensory neuropeptide; inflammatory mediator
Manufacturer:  Tocris Bioscience™ 1156/5
Catalog No. 11565
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Sensory neuropeptide and inflammatory mediator. Exerts excitatory effects on central and peripheral neurons, constricts bronchioles and is involved in pain transmission.
Specifications
Specifications
5mg