Learn More
Tocris Bioscience™ TAT 14 Available on GSA/VA Contract for Federal Government customers only.
Nrf2 pathway activator; blocks Nrf2/Keap1 interaction
Manufacturer:  Tocris Bioscience™ 4811/1
Catalog No. 48-111
Add to cart
This item is not returnable. View return policy
Description
Description
Nrf2 activator; inhibits Nrf2/Keap1 interaction. Induces upregulation of Nrf2 pathway downstream gene expression including heme-oxygenase 1. Suppresses LPS-induced TNF-α expression in THP-1 cells.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
1mg