Learn More
Tocris Bioscience™ TT 232 Available on GSA/VA Contract for Federal Government customers only.
sst1/sst4 somatostatin receptor agonist
Manufacturer:  Tocris Bioscience™ 4639/1
Catalog No. 46-391
Add to cart
This item is not returnable. View return policy
Description
Description
Peptide agonist for sst1/sst4 somatostatin receptors. Somatostatin analog; inhibits tyrosine kinase activity in human colon tumor cell lines. Demonstrates an antiproliferative effect both in vitro and in vivo and induces apoptosis in a pancreatic tumor cell line. Does not inhibit growth hormone release or secretion.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
1mg