Learn More
Tocris Bioscience™ ZIP
Cell-permeable inhibitor of atypical PKC isozyme PKMζ
Supplier:  Tocris Bioscience™ 2549/1
Catalog No. 25-491
Add to Cart
This item is not returnable. View return policy
Description
Description
ZIP is a novel, cell-permeable inhibitor of protein kinase Mζ (PKMζ), a constitutively active, atypical PKC isozyme involved in LTP maintenance. Selectively blocks PKMζ-induced synaptic potentiation in hippocampal slices in vitro. Reverses late-phase LTP (IC50 = 1 - 2.5 μM) and produces persistent loss of 1-day-old spatial memory following central administration in vivo.
Control Peptide and Biotinylated Peptide also available.
Specifications
Specifications
observed_molecular_weight: 1928.4
1 mg