Learn More
Tocris™ Tocriscreen Epigenetics Library
A library of 101 epigenetic compounds (250 μL, 10 mM DMSO solutions)
Manufacturer:  Tocris™ 6801/1 SET
Catalog No. 68-011-SET
Add to cart
This item is not returnable. View return policy
Description
Description
Tocriscreen Epigenetics Library is a library of 101 epigenetic compounds (250 μL, 10 mM solutions). Contains compounds covering >40 epigenetic targets including epigenetic readers, writers, erasers and transcriptional modulators. Includes the latest chemical tools to target p300/CBP, PCAF, BRPF1 and SMYD2.
This library replaces the Tocriscreen Epigenetics Toolbox (Cat. No. 5268).
If this library does not suit your needs, please submit your requirements through our Tocriscreen PRO custom compound library service .
Specifications
Specifications
A library of 101 epigenetic compounds (250 μL, 10 mM DMSO solutions)
1 Set
Safety and Handling
Safety and Handling
Recommended Storage : Store at -20°C