Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocrisolve 100, Tocris

Water-soluble emulsion; negative control for reagents in Tocrisolve™ 100

Manufacturer:  R&D Systems 1684/10MIL

Quantity 10mL

 View more versions of this product

Catalog No. 16-841-0MIL


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

>98%
Tocrisolve™ 100

Specifications

>98%
10mL
Tocrisolve™ 100

Safety and Handling

Recommended Storage : Store at +4°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit