Learn More
Thermo Scientific™ Todd Hewitt Broth Available on GSA/VA Contract for Federal Government customers only.
Reduce media prep time while cultivating beta hemolytic streptococci and encouraging the production of antigenic streptococci hemolysins with pre-filled, ready-to-use tubes.
$148.26 - $890.40
Specifications
Description Todd Hewitt Broth
Manufacturing Origin United States
View More Specs
Products 4
Catalog Number Mfr. No. Format Quantity Fill Volume Price Quantity & Availability  
Catalog Number Mfr. No. Format Quantity Fill Volume Price Quantity & Availability  
R064802 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
R064802
15mm x 103mm Tube 20/Pk. 0.5 mL Pack of 20 for $196.77
Add to cart
R064804
Accessories View Documents
Thermo Scientific™
R064804
Tube 100/Pk. 5 mL Case of 100 for $890.40
Add to cart
R064805 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
R064805
15mm x 103mm Tube 20/Pk. 5 mL Pack of 20 for $179.55
Add to cart
R064808
Accessories View Documents
Thermo Scientific™
R064808
15mm x 103mm Tube 20/Pk. 3 mL Pack of 20 for $148.26
Add to cart
Description
Description
Cultivate beta hemolytic streptococci, encourage the production of antigenic streptococci hemolysins, and isolate pathogenic microorganisms using convenient, pre-filled, ready-to-use Thermo Scientific™ Todd Hewitt Broth.
Remove the need for pipetting, dispensing or autoclaving with convenient, pre-filled and ready-to-use tubes of Todd Hewitt Broth, available in multiple formats and formulations.
Streptococci are facultatively anaerobic, catalase-negative, gram-positive cocci that occur as pairs or chains. They cause a wide variety of clinical diseases including pharyngitis, scarlet fever, otitis media, impetigo, meningitis, neonatal sepis, pneumonia, endocarditis, osteomyelitis, and necrotizing fascitis. One of the most useful phenotypic characteristics of streptococci is the hemolytic reaction, generally classified as alpha, beta or gamma according to the appearance of zones around colonies growing on Blood Agar. Beta-hemolytic streptococci are further characterized by Lancefield groups based on the antigenic differences in group-specific polysaccharides located in the bacterial cell wall.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Todd Hewitt Broth
United States