Learn More
Selleck Chemical LLCSupplier Diversity Partner Tofacitinib (CP-690550) Citrate 50mg 540737-29-9  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S500150MG
Tofacitinib citrate (CP-690550 citrate) is a novel inhibitor of JAK with IC50 of 1 nM, 20 nM and 112 nM against JAK3, JAK2, and JAK1, respectively. Tofacitinib citrate has anti-infection activity.
Catalog No. 50-698-8