Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tollip Antibody (SB40a), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP128621PECY55
Description
Tollip Monoclonal specifically detects Tollip in Human samples. It is validated for Flow Cytometry.Specifications
Tollip | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
TOLLIP | |
Recombinant Tollip | |
0.1 mL | |
Primary | |
Human Tollip; Does not react to TLR2, TLR4, TLR6, TLR7, TLR8 and TLR10 recombinant proteins | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
SB40a | |
Flow Cytometry | |
adapter protein, FLJ33531, IL-1RAcPIP, toll interacting protein, toll-interacting protein | |
Mouse | |
Protein A or G purified | |
Signal Transduction | |
54472.0 | |
Human | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction