Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tolmetin Sodium Salt, Dihydrate, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
64490-92-2
C15H18NNaO5
315.30
QQILXENAYPUNEA-UHFFFAOYSA-M
sodium 2-[1-methyl-5-(4-methylbenzoyl)-1H-pyrrol-2-yl]acetate dihydrate
O.O.[Na+].CN1C(CC([O-])=O)=CC=C1C(=O)C1=CC=C(C)C=C1
64490-92-2
C15H18NNaO5
315.30
QQILXENAYPUNEA-UHFFFAOYSA-M
sodium 2-[1-methyl-5-(4-methylbenzoyl)-1H-pyrrol-2-yl]acetate dihydrate
O.O.[Na+].CN1C(CC([O-])=O)=CC=C1C(=O)C1=CC=C(C)C=C1

T3958, 64490-92-2, C15H14NO3Na•2H2O

Spectrum™ Tolmetin Sodium Salt, Dihydrate, USP is a nonsteroidal anti-inflammatory drug used to reduce pain, swelling, and joint stiffness from rheumatoid arthritis and osteoarthritis. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Loss on Drying 10.4-12.4%
Quantity 25 g
CAS Max % 1
Percent Purity 98 to 102%
Grade USP