Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tolnaftate, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
1 g
10 g
50 g
250 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
2398-96-1
C19H17NOS
307.41
FUSNMLFNXJSCDI-UHFFFAOYSA-N
N-methyl-N-(3-methylphenyl)-1-(naphthalen-2-yloxy)methanethioamide
CN(C(=S)OC1=CC=C2C=CC=CC2=C1)C1=CC=CC(C)=C1
2398-96-1
C19H17NOS
307.41
FUSNMLFNXJSCDI-UHFFFAOYSA-N
N-methyl-N-(3-methylphenyl)-1-(naphthalen-2-yloxy)methanethioamide
CN(C(=S)OC1=CC=C2C=CC=CC2=C1)C1=CC=CC(C)=C1

TO102, 2398-96-1, C19H17NOS

Spectrum™ Tolnaftate, USP is an antifungal medication that fights infections caused by fungus such as athlete's foot and jock itch. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Poly Pail
Ignition Residue 0.001
Melting Range 110-113°C
Loss on Drying 0.005
Quantity 1 kg
CAS Max % 1
Percent Purity 98 to 102%
Grade USP