Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tolu Balsam, USP, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
Packaging:
Amber Glass Bottle
Poly Pail
9000-64-0
9000-64-0

TO101, 9000-64-0

Spectrum™ Tolu Balsam, USP is applied directly to the skin to treat bedsores, cracked nipples, cracked lips, and minor skin cuts. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Poly Pail
Quantity 2.5 kg
Saponification Value 154-220
CAS Max % 1
Grade USP