Alfa Aesar™ Toluene-2,4-diisocyanate, tech. 80%, remainder 2,6-diisocyanate

Manufacturer: Alfa Aesar™ 36733A1

Quantity 1kg

 View more versions of this product

Catalog No. AA36733A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 584-84-9
    Formula Weight 174.16
    MDL Number MFCD00002011
    UN Number UN2078
    Beilstein 744602
    Merck Index 14,9530
    Synonym 4-Methyl-m-phenylene diisocyante; Tolylene-2,4-diisocyanate
    Chemical Name or Material Toluene-2,4-diisocyanate
    Grade Technical Grade

    Specifications

    Formula Weight 174.16
    Health Hazard 2 GHS H Statement
    H330-H334-H351-H315-H319-H317-H335-H204
    Fatal if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Suspected of causing cancer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    Fire or projection hazard.
    Health Hazard 3 GHS P Statement
    P260-P304+P340-P305+P351+P338-P320-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00002011
    Beilstein 744602
    Sensitivity Moisture Sensitive
    Synonym 4-Methyl-m-phenylene diisocyante; Tolylene-2,4-diisocyanate
    Health Hazard 1 Danger
    Melting Point 19.5° to 21.5°C
    RTECS Number CZ6300000
    TSCA TSCA
    Physical Form Liquid
    CAS 584-84-9
    Refractive Index 1.567
    Color Colorless
    Quantity 1kg
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOLUENE DIISOCYANATE
    UN Number UN2078
    Merck Index 14,9530
    Density 1.225g/mL
    Boiling Point 251°C
    Flash Point 132° (270°F)
    Chemical Name or Material Toluene-2,4-diisocyanate
    Assay 80%, Remainder 2,6-diisocyanate
    Grade Technical Grade

    GHS H Statement
    H330-H334-H351-H315-H319-H317-H335-H204
    Fatal if inhaled.
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    Suspected of causing cancer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    May cause respiratory irritation.
    Fire or projection hazard.

    GHS P Statement
    P260-P304+P340-P305+P351+P338-P320-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit