Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Toluene, 99.7% min, HPLC Grade, Alfa Aesar™
Click to view available options
Quantity:
1L
4L
4 x 1L
Specifications
Assay Percent Range | 99.7% min. |
Beilstein | 635760 |
Solubility Information | Miscible with ethanol,benzene,diethyl ether,acetone,chloroform,glacial acetic acid and carbon disulfide. Immiscible with water. |
Formula Weight | 92.14 |
Physical Form | Liquid |
Grade | HPLC |
Density | 0.867 |
Refractive Index | 1.4967 |
Flash Point | 4°C (40°F) |
Boiling Point | 111°C |
Show More |
Safety and Handling
P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H336-H361d-H373
DOTInformation : Hazard Class: 3; Packaging Group: II
EINECSNumber : 203-625-9
RTECSNumber : XS5250000
TSCA : Yes
Recommended Storage : Ambient temperatures