Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, 99.85%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
1L
2.5L
108-88-3
C7H8
92.141
MFCD00008512
YXFVVABEGXRONW-UHFFFAOYSA-N
methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene
1140
CHEBI:17578
toluene
CC1=CC=CC=C1
108-88-3
C7H8
92.141
MFCD00008512
YXFVVABEGXRONW-UHFFFAOYSA-N
methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene
1140
CHEBI:17578
toluene
CC1=CC=CC=C1
Additional Information Vapor Pressure: 29mbar at 20°C
Assay Percent Range 99.85%
Linear Formula C6H5CH3
Beilstein 05, 280
Merck Index 15, 9688
Solubility Information Solubility in water: almost insoluble (0.5g/L at 20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, acetone,, ether, petroleum ether, benzene, ch2cl2, dmf, gla, cial acetic acid, chcl3, ccl4.
Formula Weight 92.14
Physical Form Liquid
Percent Purity 99.85%
Grade Extra Dry over Molecular Sieve
Density 0.8660g/mL
Hydrochloric Acid 0.002 % max.
Infrared Spectrum Authentic
Refractive Index 1.4968
Solubility Solubility in water: almost insoluble (0.5g/L at 20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, acetone,, ether, petroleum ether, benzene, ch2cl2, dmf, gla, cial acetic acid, chcl3, ccl4.
Specific Gravity 0.866
Water 0.005% Max. (Coulometric)
Flash Point 4°C
Viscosity 0.6 mPa.s (20°C)
Boiling Point 111.0°C
Color Colorless
Melting Point -95.0°C
Quantity 500mL
Chemical Name or Material Toluene, water <50 ppm
Show More Show Less

GHS H Statement
May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure if inhaled. May be fatal if swallowed and enters airways. Causes skin irritation. Suspected of damaging the unborn child. Highly flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection.

Danger