Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, 99+%, extra pure, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
5L
10L
25L
Packaging:
Glass Bottle
Metal Drum
Plastic drum
108-88-3
C7H8
92.141
MFCD00008512
YXFVVABEGXRONW-UHFFFAOYSA-N
methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene
1140
CHEBI:17578
toluene
CC1=CC=CC=C1
108-88-3
C7H8
92.141
MFCD00008512
YXFVVABEGXRONW-UHFFFAOYSA-N
methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene
1140
CHEBI:17578
toluene
CC1=CC=CC=C1
Additional Information Vapor Pressure: 29mbar at 20°C
Assay Percent Range 99+%
Linear Formula C6H5CH3
Beilstein 05, 280
Merck Index 15, 9688
Solubility Information Solubility in water: almost insoluble (0.5g/L at 20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, acetone,, ether, petroleum ether, benzene, ch2cl2, dmf, gla, cial acetic acid, chcl3, ccl4.
Formula Weight 92.14
Physical Form Liquid
Percent Purity ≥99%
Grade Extra Pure
Density 0.8660g/mL
Packaging Metal Drum
Refractive Index 1.4968
Residue after Evaporation 0.003% max.
Specific Gravity 0.866
Water 0.05% max. (K.F.)
Flash Point 4°C
Viscosity 0.6 mPa.s (20°C)
Boiling Point 111.0°C
Color Undesignated
Melting Point -95.0°C
Quantity 25L
Chemical Name or Material Toluene
Show More Show Less

GHS P Statement May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure if inhaled. May be fatal if swallowed and enters airways. Causes skin irritation. Suspected of damag

Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0

Signal Word: DANGER!