Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, 99+%, for spectroscopy, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
Packaging:
Glass bottle
108-88-3
C7H8
92.14
MFCD00008512
YXFVVABEGXRONW-UHFFFAOYSA-N
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
1140
CHEBI:17578
toluene
CC1=CC=CC=C1
108-88-3
C7H8
92.14
MFCD00008512
YXFVVABEGXRONW-UHFFFAOYSA-N
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
1140
CHEBI:17578
toluene
CC1=CC=CC=C1
For UV/VIS spectroscopy
Absorbance (1 cm cell vs water), 0.01 max. at 350nm, 0.02 max. at 335nm, 0.07 max. at 310nm, 0.1 max. at 300nm, 0.4 max. at 288nm, 1 max. at 285nm
Absorbance Conditions (1 cm cell vs water)
Additional Information Vapor Pressure: 29mbar at 20°C
Assay Percent Range 99+%
Linear Formula C6H5CH3
Beilstein 05, 280
Merck Index 15, 9688
Formula Weight 92.14
Physical Form Liquid
Percent Purity ≥99.0%
Grade Spectroscopy
Density 0.8660g/mL
Packaging Glass bottle
Residue after Evaporation 0.001% max.
Water 0.03% max.
Color Undesignated
Quantity 1L
Chemical Name or Material Toluene, 99+%
Show More Show Less

GHS H Statement:
May cause drowsiness or dizziness.
May cause damage to organs through prolonged or repeated exposure.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Suspected of damaging the unborn child.
Highly flammable liquid and vapor.

Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0

Signal Word: DANGER!

EINECSNumber : 203-625-9

RTECSNumber : XS5250000

TSCA : TSCA