Learn More
Toluene, ACS, 99.5% min, Thermo Scientific Chemicals
Description
Toluene is used as solvent for paints, paint thinners, silicone sealants, many chemical reactants, rubber, printing ink, adhesives (glues), lacquers, leather tanners and disinfectants. It is an important raw material to synthesize pharmaceuticals, paints, resins, dyes, toluene diisocyanate and explosives like trinitrotoluene. It is used as standard material for chromatography and analytical reagent. It plays an important role in the manufacture of benzene, xylene, phenol, poly urethane polymers, cosmetics and jet fuel blends. It is used as fuel additive to increase octane ratings and a coolant for its good heat transfer capabilities in sodium cold traps used in nuclear reactor system loops.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | °C to 93°C |
Color | Colorless |
Appearance | Colorless liquid |
Density | 0.867 |
Boiling Point | 111°C |
Quantity | 1 L |
Flash Point | 4°C (40°F) |
Odor | Aromatic |
Physical Form | Liquid |
Refractive Index | 1.4967 |
Show More |
Safety and Handling
Hazard Category | H225-H304-H315-H319-H336-H361d-H373 |
Precautionary Statement | P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c |
DOTInformation | DOT Class: 3; Packing Group: II |
EINECSNumber | 203-625-9 |
RTECSNumber | XS5250000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.