Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, anhydrous, 99.8%, Alfa Aesar™

Click to view available options
Quantity:
250mL
1L
4L
108-88-3
C7H8
92.141
MFCD00008512
YXFVVABEGXRONW-UHFFFAOYSA-N
methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene
1140
CHEBI:17578
toluene
CC1=CC=CC=C1
108-88-3
C7H8
92.141
MFCD00008512
YXFVVABEGXRONW-UHFFFAOYSA-N
methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene
1140
CHEBI:17578
toluene
CC1=CC=CC=C1
Assay Percent Range 99.8%
Beilstein 635760
Solubility Information Miscible with ethanol,alcohol,benzene,diethyl ether,acetone,chloroform,glacial acetic acid and carbon disulfide. Immiscible with water.
Formula Weight 92.14
Physical Form Liquid
Density 0.867 g/mL
Refractive Index 1.4967
Flash Point 4°C (40°F)
Boiling Point 111°C
Melting Point °C to 93°C
Quantity 250mL
Chemical Name or Material Toluene, anhydrous
Show More Show Less

P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c

H225-H304-H315-H319-H336-H361d-H373

DOTInformation : Hazard Class: 3; Packaging Group: II

EINECSNumber : 203-625-9

RTECSNumber : XS5250000

TSCA : Yes

Recommended Storage : Ambient temperatures