Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, Anhydrous, septum-sealed, for applications requiring low water content, >99.9%, Honeywell Burdick & Jackson™

Click to view available options
Quantity:
6 x 100 mL
6 x 1 L
Packaging:
Septum-sealed Bottle
108-88-3
C7H8
92.14
MFCD00008512
YXFVVABEGXRONW-UHFFFAOYSA-N
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
1140
CHEBI:17578
toluene
CC1=CC=CC=C1
108-88-3
C7H8
92.14
MFCD00008512
YXFVVABEGXRONW-UHFFFAOYSA-N
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
1140
CHEBI:17578
toluene
CC1=CC=CC=C1

Anhydrous, septum-sealed, for applications requiring low water content, >99.9%

  • Water content ranging from <10 ppm to <50 ppm
  • Low residue
  • Convenient and effective packaging ensures product quality
  • Certificate of Analysis printed on the label

 

CAS Max % 100.0000%
UN Number UN1294
Beilstein 635760
Formula Weight 92.14
Percent Purity >99.9%
Density 0.867 g/cm3
Boiling Point 110.6°C
Quantity 6 x 1 L
Flash Point 4°C
Packaging Septum-sealed Bottle
Physical Form anhydrous
Chemical Name or Material Toluene, Anhydrous, septum-sealed, for applications requiring low water content, >99.9%
Show More Show Less

ShelfLife : 365 days from date of manufacture

DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Toluene