Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Toluene, Anhydrous, septum-sealed, for applications requiring low water content, >99.9%, Honeywell Burdick & Jackson™
Click to view available options
Quantity:
6 x 100 mL
6 x 1 L
Packaging:
Septum-sealed Bottle
Description
- Water content ranging from <10 ppm to <50 ppm
- Low residue
- Convenient and effective packaging ensures product quality
- Certificate of Analysis printed on the label
Specifications
CAS Max % | 100.0000% |
UN Number | UN1294 |
Beilstein | 635760 |
Formula Weight | 92.14 |
Percent Purity | >99.9% |
Density | 0.867 g/cm3 |
Boiling Point | 110.6°C |
Quantity | 6 x 100 mL |
Flash Point | 4°C |
Packaging | Septum-sealed Bottle |
Show More |
Safety and Handling
ShelfLife | 365 days from date of manufacture |
DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Toluene |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction