Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
1L
4L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle

Chemical Identifiers

108-88-3
C7H8
92.141
YXFVVABEGXRONW-UHFFFAOYSA-N
1140
CHEBI:17578

High quality chemicals for laboratory and specialized industrial use

    For Laboratory,Research,or Manufacturing Use
SMILES CC1=CC=CC=C1
IUPAC Name toluene
Formula Weight 92.14
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.86kg
Flash Point 4.00°C
Quantity 4L
Chemical Name or Material Toluene
Grade Baker Analyzed™ ACS Reagent, Meets ACS reagent chemical requirements