Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
1L
4L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle
108-88-3
C7H8
92.14
MFCD00008512
YXFVVABEGXRONW-UHFFFAOYSA-N
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
1140
CHEBI:17578
toluene
CC1=CC=CC=C1
108-88-3
C7H8
92.14
MFCD00008512
YXFVVABEGXRONW-UHFFFAOYSA-N
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
1140
CHEBI:17578
toluene
CC1=CC=CC=C1

High quality chemicals for laboratory and specialized industrial use

    For Laboratory,Research,or Manufacturing Use
Formula Weight 92.14
Grade Meets ACS reagent chemical requirements
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.86kg
Flash Point 4.00°C
Quantity 500mL
Chemical Name or Material Toluene