Alfa Aesar™ Toluene-d8, 100% (Isotopic)

Manufacturer: Alfa Aesar™ 42356SA

Quantity 2 each

 View more versions of this product

Catalog No. AA42356SA

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2037-26-5
    Formula Weight 100.19
    MDL Number MFCD00044638
    UN Number UN1294
    Chemical Name or Material Toluene-d8, Isotopic

    Specifications

    Formula Weight 100.19
    Packaging Prescored Ampules
    Health Hazard 2 GHS H Statement
    H225-H304-H361-H373-H315-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes skin irritation.
    May cause drowsiness or dizziness.
    Health Hazard 3 GHS P Statement
    P210-P260-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00044638
    Density 0.94g/mL
    Physical Form Liquid
    Quantity 2 each
    Flash Point 10°C (50°F)
    Boiling Point 111°C
    Melting Point −93°C
    CAS 2037-26-5
    UN Number UN1294
    Health Hazard 1 Danger
    Odor Aromatic
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TOLUENE
    Chemical Name or Material Toluene-d8, Isotopic
    Assay 100%

    GHS H Statement
    H225-H304-H361-H373-H315-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes skin irritation.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P260-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit