Learn More
Toluene-d8, 99.6%(Isotopic), Thermo Scientific Chemicals
Description
Toluene is mainly used as a precursor to benzene. Paints, paint thinners, silicone sealants, many chemical reactants, rubber, printing ink, adhesives (glues), lacquers, and leather tanners. And also used as an octane booster in gasoline fuels used in internal combustion engines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Physical Form | Liquid |
Quantity | 10 g |
Density | 0.94 g/mL |
Flash Point | 10°C (50°F) |
Odor | Aromatic |
Melting Point | -93°C |
Boiling Point | 111°C |
UN Number | UN1294 |
Solubility Information | Soluble in water.0.5g/L (20°C) |
Formula Weight | 100.19 |
Show More |
Safety and Handling
Hazard Category | H225-H304-H315-H319-H336-H361d-H373 |
Hazard Statement | GHS H Statement H225-H304-H361-H373-H315-H336 Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Causes skin irritation. May cause drowsiness or dizziness. |
Precautionary Statement | P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280i-P281-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P331-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TOLUENE |
EINECSNumber | 218-009-5 |
TSCA | No |
Recommended Storage | Ambient |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.