Alfa Aesar™ Toluene-d8, 99.6%(Isotopic)

Manufacturer: Alfa Aesar™ 3647703

Quantity 1g

 View more versions of this product

Catalog No. AA3647703

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 2037-26-5
    Formula Weight 100.19
    MDL Number MFCD00044638
    UN Number UN1294
    Chemical Name or Material Toluene-d8, Isotopic

    Specifications

    Formula Weight 100.19
    Health Hazard 2 GHS H Statement
    H225-H304-H361-H373-H315-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes skin irritation.
    May cause drowsiness or dizziness.
    Health Hazard 3 GHS P Statement
    P210-P260-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN1294
    Density 0.94g/mL
    Physical Form Liquid
    Flash Point 10° (50°F)
    MDL Number MFCD00044638
    Health Hazard 1 Danger
    Odor Aromatic
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TOLUENE
    Boiling Point 111°C
    Melting Point −93°C
    Quantity 1g
    CAS 2037-26-5
    Chemical Name or Material Toluene-d8, Isotopic
    Assay 99.6%

    GHS H Statement
    H225-H304-H361-H373-H315-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Suspected of damaging fertility or the unborn child.
    May cause damage to organs through prolonged or repeated exposure.
    Causes skin irritation.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P260-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit