Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, Environmental Grade, >99.8%, Alfa Aesar™

Suitable For Pesticide Residue Analysis

Manufacturer:  Alfa Aesar 43061M6

Quantity 4 x 4L

 View more versions of this product

Catalog No. AA43061M6


 

Due to product restrictions, please to purchase or view availability for this product.


Specifications Description & Specifications

Chemical Identifiers

108-88-3
>99.8%
92.14
Toluene, Submicron filtered
Liquid
C6H5CH3
MFCD00008512
Environmental Grade

Specifications

108-88-3
>99.8%
92.14
0.867 g/cm3 at 20°C
Aromatic
Clear colorless
Toluene, Submicron filtered
Liquid
C6H5CH3
MFCD00008512
535°C (995°F)
22mm Hg at 20°C
4 x 4L
Environmental Grade

Safety and Handling

SDS & Safety Documents

H225-H304-H361-H373-H315-H336: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Causes skin irritation. May cause drowsiness or dizziness.

DOTInformation : DOT Class: 3, Packaging Group: II

EINECSNumber : 203-625-9

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit