Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, HPLC Grade, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Glass Bottle
108-88-3
C7H8
92.141
YXFVVABEGXRONW-UHFFFAOYSA-N
methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene
1140
CHEBI:17578
toluene
CC1=CC=CC=C1
108-88-3
C7H8
92.141
YXFVVABEGXRONW-UHFFFAOYSA-N
methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene
1140
CHEBI:17578
toluene
CC1=CC=CC=C1

High purity solvents for use in analytical and preparative separations

    For use in Liquid Chromatography (HPLC & UHPLC) & Spectrophotometry; For Laboratory, Research, or Manufacturing Use
Formula Weight 92.14
Grade HPLC
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.86kg
Flash Point 4.00°C
Quantity 1L
Chemical Name or Material Toluene