Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene (Laboratory), Fisher Chemical™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
4L
20L
Packaging:
Glass Bottle
Steel Pail
108-88-3
C7H8
92.141
MFCD00008512
YXFVVABEGXRONW-UHFFFAOYSA-N
methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene
1140
CHEBI:17578
toluene
CC1=CC=CC=C1
108-88-3
C7H8
92.141
MFCD00008512
YXFVVABEGXRONW-UHFFFAOYSA-N
methylbenzene,toluol,phenylmethane,benzene, methyl,methylbenzol,methacide,toluen,antisal 1a,tolu-sol,monomethyl benzene
1140
CHEBI:17578
toluene
CC1=CC=CC=C1

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Additional Information Vapor Pressure: 29mbar at 20°C
Linear Formula C6H5CH3
Formula Weight 92.14
Physical Form Liquid
Grade Laboratory
Density 0.866g/cm³
Identification Pass Test
Packaging Glass Bottle
Vapor Pressure 29mbar at 20°C
Viscosity 0.6 mPa/s at 20°C
Boiling Point 111°C
Color Colorless
Melting Point -95°C
Quantity 4L
Chemical Name or Material Toluene
Show More Show Less

DANGER!

Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0

Signal Word: DANGER!

DOTInformation : DOT Class 3, : Flammable Liquid