Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, Low water, BakerDRY™, J.T.Baker™

Click to view available options
Quantity:
100mL
1L
Packaging:
Glass Bottle
Narrow Mouth Amber Glass Bottle, w/ Septum

Chemical Identifiers

108-88-3
C7H8
92.141
YXFVVABEGXRONW-UHFFFAOYSA-N
1140
CHEBI:17578

Specifically manufactured for use in organometallic and oligonucleotide synthesis

CAS 108-88-3
Molecular Formula C7H8
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
SMILES CC1=CC=CC=C1
IUPAC Name toluene
Molecular Weight (g/mol) 92.141
PubChem CID 1140
ChEBI CHEBI:17578
Formula Weight 92.14
Packaging Glass Bottle
Specific Gravity 1L = 0.86kg
Flash Point 4.00°C
Quantity 100mL
Chemical Name or Material Toluene, Low water
Grade BakerDRY™
Show More Show Less