Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, Puriss. p.a., ACS Reagent, ≥99.7% (GC), Honeywell Riedel-de Haën™

Click to view available options
Quantity:
25 mL
4 x 25 mL
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Stainless Steel Can
108-88-3
C7H8
92.14
MFCD00008512
YXFVVABEGXRONW-UHFFFAOYSA-N
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
1140
CHEBI:17578
toluene
CC1=CC=CC=C1
108-88-3
C7H8
92.14
MFCD00008512
YXFVVABEGXRONW-UHFFFAOYSA-N
methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
1140
CHEBI:17578
toluene
CC1=CC=CC=C1

Puriss. p.a., ACS Reagent,≥99.7% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
Melting Point -93°C (literature)
CAS Max % 100.0000%
Linear Formula C6H5CH3
UN Number UN1294
Beilstein 635760
Formula Weight 92.14
Percent Purity >99.7%
Grade ACS Reagent
Density 0.865 g/mL at 25 °C(lit.)
Boiling Point 110.6°C
Quantity 4 x 25 mL
Flash Point 4°C
Packaging Glass Bottle
Chemical Name or Material Toluene, Puriss. p.a., ACS Reagent, ≥99.7% (GC)
Show More Show Less

ShelfLife : 1260 days from date of manufacture

DOTInformation : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Toluene

EINECSNumber : 203-625-9

Recommended Storage : Room Temp