Learn More


Toluene (Spectranalyzed™), Fisher Chemical
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Absorbance | Pass Test |
Additional Information | Vapor Pressure: 29mbar at 20°C |
Assay Percent Range | ≥99.5 % |
Linear Formula | C6H5CH3 |
Formula Weight | 92.14 |
Physical Form | Liquid |
Percent Purity | ≥99.5% |
Grade | Also meets ACS specifications |
Copper (Cu) | 0.1ppm max. |
Density | 0.866g/cm³ |
Show More |
Safety and Handling


DANGER!
Emergency Overview
Flammable liquid and vapor. Causes eye, skin, and respiratory tract irritation. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Danger of serious damage to health by prolonged exposure. Possible risk of harm to the unborn child. May cause adverse kidney effects. May cause adverse liver effects.
NFPA
Health:2 Flammability:3 Instability:0
Signal Word: DANGER!