Toluene, Spectrophotometric Grade, >99.7%, Alfa Aesar™

Maximum level of impurities: Evaporation residue 4ppm, H2O 0.03%

Manufacturer: Alfa Aesar 19376M6

Quantity 4 x 4L

 View more versions of this product

Catalog No. AA19376M6

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 108-88-3
    Physical Form Liquid
    Assay Percent Range >99.7%
    Molecular Formula C6H5CH3
    Formula Weight 92.14
    MDL Number MFCD00008512
    Chemical Name or Material Toluene
    Grade Spectrophotometric grade

    Specifications

    Physical Form Liquid
    Color Clear colorless
    Formula Weight 92.14
    Quantity 4 x 4L
    DOT Information Hazard Class: 3, Packing Group: II
    EINECS Number 203-625-9
    Impurities Maximum level of impurities: Evaporation residue 4ppm, H2O 0.03%
    Odor Aromatic
    Appearance Colorless liquid
    Assay Percent Range >99.7%
    Molecular Formula C6H5CH3
    MDL Number MFCD00008512
    CAS 108-88-3
    Health Hazard 1 H225-H304-H361-H373-H315-H336: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Causes skin irritation. May cause drowsiness or dizziness.
    Health Hazard 2 Wear suitable protective clothing and gloves. If swallowed seek medical advice immediately and show this container or label. If swallowed, do not induce vomiting: seek medical advice immediately & show this container or label.
    Chemical Name or Material Toluene
    Grade Spectrophotometric grade

    Wear suitable protective clothing and gloves. If swallowed seek medical advice immediately and show this container or label. If swallowed, do not induce vomiting: seek medical advice immediately & show this container or label.

    H225-H304-H361-H373-H315-H336: Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. Suspected of damaging fertility or the unborn child. May cause damage to organs through prolonged or repeated exposure. Causes skin irritation. May cause drowsiness or dizziness.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit