Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Toluene)tricarbonylchromium, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass Bottle
12083-24-8
C10H8CrO3
228.167
MFCD00010583
KEXFCQUTYPQKCG-UHFFFAOYSA-N
toluene tricarbonylchromium 0,chromium, tricarbonyl 1,2,3,4,5,6-eta-methylbenzene-9ci,tris carbon monoxide toluene chromium
11172208
carbon monoxide;chromium;toluene
CC1=CC=CC=C1.[C-]#[O+].[C-]#[O+].[C-]#[O+].[Cr]
12083-24-8
C10H8CrO3
228.167
MFCD00010583
KEXFCQUTYPQKCG-UHFFFAOYSA-N
toluene tricarbonylchromium 0,chromium, tricarbonyl 1,2,3,4,5,6-eta-methylbenzene-9ci,tris carbon monoxide toluene chromium
11172208
carbon monoxide;chromium;toluene
CC1=CC=CC=C1.[C-]#[O+].[C-]#[O+].[C-]#[O+].[Cr]
Infrared Spectrum Authentic
Elemental Analysis 97.5% min.
Quantity 1g
Flash Point 76°C
Color Orange-Yellow to Yellow
Packaging Glass Bottle
Melting Point 78.0°C to 82.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 228.17
Physical Form Crystalline Powder or Crystals
Percent Purity 98%
Chemical Name or Material (Toluene)tricarbonylchromium
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Flammable solid.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger