Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluidine Blue, 1% (w/v) Aqueous Solution, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
4 L
Packaging:
Poly Bottle
92-31-9
C15H16ClN3S
305.82
HNONEKILPDHFOL-UHFFFAOYSA-M
3-amino-7-(dimethylamino)-2-methyl-5λ⁴-phenothiazin-5-ylium chloride
[Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1
92-31-9
C15H16ClN3S
305.82
HNONEKILPDHFOL-UHFFFAOYSA-M
3-amino-7-(dimethylamino)-2-methyl-5λ⁴-phenothiazin-5-ylium chloride
[Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=[S+]C2=C1

T-140, 92-31-9

Spectrum™ Toluidine Blue, 1 Percent (w/v) Aqueous Solution is a blue cationic (basic) dye used in histology to test for lignin and for staining semi-thin (0.5 to 1 μm) sections of resin-embedded tissue. Spectrum solutions utilize the highest quality raw materials appropriate to your product. Most raw materials meet or exceed the specifications established by the American Chemical Society. Where appropriate, many finished products are traceable to NIST Standard Reference Materials. Manufacturing, quality control testing, and packaging are all performed in Spectrum’s own facilities. Record keeping and sample retention of all produced lots ensure product consistency and complete traceability.

CAS Max % 1
Grade Chemical Solution
Packaging Poly Bottle
Quantity 4 L