Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tolylene 2,6-diisocyanate, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
91-08-7
C9H6N2O2
174.16
MFCD00002010
RUELTTOHQODFPA-UHFFFAOYSA-N
2,6-diisocyanatotoluene, toluene-2,6-diisocyanate, 2,6-toluene diisocyanate, 2-methyl-m-phenylene diisocyanate, 2,6-tdi, m-tolylene diisocyanate, meta-tolylene diisocyanate, toluene 2,6-diisocyanate, benzene, 1,3-diisocyanato-2-methyl, 2,6-diisocyanato-1-methylbenzene
7040
CHEBI:53557
1,3-diisocyanato-2-methylbenzene
CC1=C(C=CC=C1N=C=O)N=C=O
91-08-7
C9H6N2O2
174.16
MFCD00002010
RUELTTOHQODFPA-UHFFFAOYSA-N
2,6-diisocyanatotoluene, toluene-2,6-diisocyanate, 2,6-toluene diisocyanate, 2-methyl-m-phenylene diisocyanate, 2,6-tdi, m-tolylene diisocyanate, meta-tolylene diisocyanate, toluene 2,6-diisocyanate, benzene, 1,3-diisocyanato-2-methyl, 2,6-diisocyanato-1-methylbenzene
7040
CHEBI:53557
1,3-diisocyanato-2-methylbenzene
CC1=C(C=CC=C1N=C=O)N=C=O
Flash Point >110°C
Density 1.2200g/mL
Viscosity 3 mPa.s (25°C)
Refractive Index 1.5700 to 1.5720
Packaging Glass bottle
Quantity 5g
Specific Gravity 1.22
Infrared Spectrum Authentic
Melting Point 13.0°C
Color Colorless to Yellow
Boiling Point 246.0°C to 247.0°C
Assay Percent Range 96% min. (GC)
Linear Formula CH3C6H3(NCO)2
Beilstein 13, III, 291
Formula Weight 174.16
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Tolylene 2, 6-diisocyanate, 97%
Show More Show Less

GHS H Statement
Suspected of causing cancer. May cause an allergic skin reaction. May cause respiratory irritation. Harmful to aquatic life with long lasting effects. Causes skin irritation. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Fatal if inhaled. Causes serious eye irritation.

GHS P Statement
Immediately call a POISON CENTER or doctor/physician. Avoid breathing dust/fume/gas/mist/vapors/spray. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician. Wear eye protection/face protection. Use personal protective equipment as required. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Avoid breathing dust/fume/gas/mist/vapors/spray. Avoid release to the environment. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS Signal Word: Danger

EINECSNumber : 202-039-0

RTECSNumber : CZ6310000

TSCA : TSCA