Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOM1L1 Mouse anti-Human, Clone: TOM1L1/4690, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30713610UG

 View more versions of this product

Catalog No. NB121745


Add to cart

Description

Description

TOM1L1 Monoclonal specifically detects TOM1L1 in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

TOM1L1
TOM1L1/4690
Unconjugated
10 mM PBS with 0.05% BSA
OK/KNS-CL.3, Src-activating and signaling molecule protein, SRCASMSrc activating and signaling molecule, target of myb 1-like 1, target of myb1 (chicken) homolog-like 1, target of myb1 (chicken)-like 1, target of myb1-like 1, Target of Myb-like protein 1, TOM1-like protein 1
Recombinant full-length human TOM1L1 protein
Protein A or G purified
RUO
Primary
10040
Immunohistochemistry (Paraffin)
0.2 mg/ml
Immunohistochemistry-Paraffin 1-2 ug/ml
Purified
Mouse
IgG
100 μg
Store at 4°C.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit