Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOM70 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA146188

Catalog No. PA146188


Only null left
Add to Cart

Description

Description

Suggested positive control: purified mitochondrial extracts and HeLa whole cell extract.

Functional mitochondria require up to 1000 proteins to function properly, with 99% synthesized as precursors in the cytoplasm and transported into the mitochondria with the aid of cytosolic chaperones and mitochondrial translocators (import components). Proteins to be imported are chaperoned to the mitochondria by the cytosolic heat shock protein (cHSP70) and are immediately pursued by Translocators of the Outer Membrane (TOMs), followed by transient interactions of the unfolded proteins with Translocators of the Inner Membrane (TIMs). TOMM70A is ubiquitously expressed in human tissues and localizes in the mitochondria. TOMM70A could play a significant role in the import of nuclear-encoded mitochondrial proteins with internal targeting sites such as ADP/ATP carriers and the uncoupling proteins.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TOM70
Polyclonal
Unconjugated
TOMM70
2610044B22Rik; D16Ium22; D16Ium22e; D16Wsu109e; FLJ90470; KIAA0719; Mitochondrial import receptor subunit TOM70; mitochondrial precursor proteins import receptor; mKIAA0719; Tom70; Tomm70; Tomm70a; Translocase of outer membrane 70 kDa subunit; translocase of outer mitochondrial membrane 70; translocase of outer mitochondrial membrane 70 homolog A; translocase of outer mitochondrial membrane 70 homolog A (S. cerevisiae); translocase of outer mitochondrial membrane 70 homolog A (yeast); translocase of outer mitochondrial membrane protein 70
Rabbit
Protein A
RUO
100352274, 28185, 304017, 9868
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with no preservative
O94826, Q75Q39, Q9CZW5
TOMM70, Tomm70a
A synthetic peptide made to a region within residues 1-100 of the human TOM70 protein.
100 μL
Primary
Human, Mouse, Rat, Rabbit
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.