Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TOMM20 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP303878100UL
Description
TOMM20 Polyclonal antibody specifically detects TOMM20 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, KnockDownSpecifications
TOMM20 | |
Unconjugated | |
Purified | |
Rabbit | |
IgG | |
100 μL | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Polyclonal | |
Human, Mouse, Rat |
Immunofluorescence, Immunohistochemistry, KnockDown, Western Blot | |
PBS with 50% glycerol, pH7.3. | |
KIAA0016Outer mitochondrial membrane receptor Tom20, MAS20, MGC117367, Mitochondrial 20 kDa outer membrane protein, mitochondrial import receptor subunit TOM20 homolog, MOM19, TOM20, translocase of outer mitochondrial membrane 20 homolog (yeast), translocase of outer mitochondrial membrane 20 homolog type II | |
Recombinant protein of human TOMM20. | |
Affinity purified | |
RUO | |
Primary | |
9804 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title