Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOMM34 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549407

Catalog No. PA549407

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The protein encoded by this gene is involved in the import of precursor proteins into mitochondria. The encoded protein has a chaperone-like activity, binding the mature portion of unfolded proteins and aiding their import into mitochondria. This protein, which is found in the cytoplasm and sometimes associated with the outer mitochondrial membrane, has a weak ATPase activity and contains 6 TPR repeats.
Specifications

Specifications

TOMM34
Polyclonal
PBS with 0.09% sodium azide
Q9CYG7
Mitochondrial import receptor subunit TOM34, hTom34, Translocase of outer membrane 34 kDa subunit, TOMM34, URCC3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
67145
Western Blot
Unconjugated
Tomm34
Liquid
TOMM34
KLH conjugated synthetic peptide between 156-188 amino acids from the Central region of human TOMM34.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit