Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOMM40L Rabbit anti-Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549376

Catalog No. PA549376


Add to cart

Description

Description

Predicted to react with bovine and mouse based on sequence homology.

Potential channel-forming protein implicated in import of protein precursors into mitochondria.
Specifications

Specifications

TOMM40L
Polyclonal
PBS with 0.09% sodium azide
A4F267
Mitochondrial import receptor subunit TOM40B, Protein TOMM40-like, TOMM40L, TOMM40B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
304971
Western Blot
Unconjugated
Tomm40l
Liquid
TOMM40L
KLH conjugated synthetic peptide between 25-58 amino acids from the N-terminal region of human TOMM40L.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit