Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TOMM5 Rabbit anti-Human, Mouse, Non-human Primate, Rat, Biotin, Polyclonal, FabGennix

Rabbit Polyclonal Antibody
Supplier: Fabgennix Inc TOM5BIOTIN

Description
TOMM5 Polyclonal antibody specifically detects TOMM5 in Human, Mouse, Non-human Primate, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western BlotSpecifications
TOMM5 | |
Polyclonal | |
Biotin | |
Tomm5 | |
1110019J04Rik; bA613M10.3; C9orf105; Mitochondrial import receptor subunit TOM5 homolog; mitochondrial outer membrane protein TOM5; Tom5; TOMM5; translocase of outer mitochondrial membrane 5; translocase of outer mitochondrial membrane 5 homolog; translocase of outer mitochondrial membrane 5 homolog (yeast) | |
Synthetic peptide taken within amino acid region 1-50 on human Mitochondrial import receptor subunit TOM5 homolog isoform 1 protein. | |
200 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.5-1.5 mg/mL | |
proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8 | |
B1AXP6, Q8N4H5 | |
Rabbit | |
Affinity chromatography | |
RUO | |
401505, 680080, 68512, 747663 | |
-20°C | |
Liquid |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title