Learn More
TOMM6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 166891AP
Catalog No. 50-173-1116
Add to cart
Description
Description
TOMM6, also named as OBTP and TOM6, is a part of the preprotein translocase complex of the outer mitochondrial membrane (TOM complex).
Specifications
Specifications
TOMM6
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q96B49, Q9CQN3
OBTP, TOM6, TOMM6
Rabbit
IgG
150 μL
-20°C
Primary
100188893, 66119, 681123
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TOMM6
Liquid
Tomm6
TOMM6 Fusion Protein Ag10037
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat