Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TOMM70A Rabbit anti-Human, Mouse, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Supplier: Abnova Corporation PAB30601
Description
Rabbit polyclonal antibody raised against partial recombinant human TOMM70A.
- High specificity
- High reactivity
Specifications
TOMM70A | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial recombinant human TOMM70A. | |
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). | |
FLJ90470 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
9868 | |
Store at 4°C for short term storage. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunofluorescence (1 - 4 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200 - 1:500) Western Blot (1:100 - 1:250) The optimal working dilution should be determined by the end user. | |
O94826 | |
TOMM70A | |
Recombinant protein corresponding to amino acids 383-512 of human TOMM70A. | |
100 μL | |
Primary | |
Human, Mouse | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction