Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tomosyn Rabbit anti-Human, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP30378120UL
Description
Tomosyn Polyclonal antibody specifically detects Tomosyn in Human samples. It is validated for Western BlotSpecifications
Tomosyn | |
Unconjugated | |
Purified | |
Rabbit | |
IgG | |
20 μL | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Polyclonal | |
Human |
Western Blot | |
PBS with 50% glycerol, pH7.3. | |
FLJ30922, Lethal(2) giant larvae protein homolog 3, LGL3, LLGL3Nbla04300, MGC141942, MGC141968, putative protein product of Nbla04300, syntaxin binding protein 5 (tomosyn), syntaxin-binding protein 5, tomosyn, tomosyn-1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 640-900 of human Tomosyn (NP_640337.3). | |
Affinity purified | |
RUO | |
Primary | |
134957 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title