Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIP™ Top Grain Goatskin Leather Protector for Novax™ Gloves

PIP™ Class 100 Static Dissipative Latex Finger Cots are ideal for ESD- sensitive areas, Defense-Avionics, Marine Electronics, Consumer Electronics, IC Industries-wire bonding, Repair Stations-assembly area

$36.59 - $60.87

Specifications

Arc Flash Class 00 Gloves
Certifications/Compliance Meets ASTM F696-06 (2011) standard
Color Natural
Description Keystone thumb; Sheared elastic back
For Use With (Application) ARC Flash
View More Specs

Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-168-2089 View Documents PIP™
1481000/7
7 Pair for $36.59
Add to cart
 
19-168-2090 View Documents PIP™
1481000/8
8 Pair for $36.59
Add to cart
 
19-168-2093 View Documents PIP™
1481000/11
11 Pair for $60.87
Add to cart
 
19-168-2094 View Documents PIP™
1481000/12
12 Pair for $36.59
Add to cart
 
Description

Description

  • Top Grain Goatskin leather construction is soft, abrasion-resistant leather recommended for applications requiring tactile sensitivity
  • Smooth
  • Provides durability and dexterity
  • Drivers gloves feature slip-on styling to provide good fit and comfort
  • Keystone thumb is ergonomic, reduces hand fatigue, offers dexterity and durability
  • Fourchettes between the fingers enhance the natural contour of the glove for a better fit
  • Elastic closure sewn inside glove for safety and comfort
  • Color coded hems for easy size identification
  • Meets ASTM F696-06 (2011) standard
Specifications

Specifications

Class 00 Gloves
Natural
ARC Flash
Driver's style
Meets ASTM F696-06 (2011) standard
Keystone thumb; Sheared elastic back
Leather
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit