Learn More
Invitrogen™ TOP10 Electrocomp™ Kit
Supplied at transformation efficiency of 1 × 109 cfu/μg supercoiled DNA for high-efficiency cloning and plasmid propagation
$299.00 - $530.00
Catalog Number | Mfr. No. | Includes | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Quantity | Price | Quantity & Availability | ||||
C66455
![]() |
Invitrogen™
C66455 |
Contains enough electrocompetent cells for 12 x 40μL rxns or 25 x 20μL rxns | 5 x 100 μL |
Each for $299.00
|
|
C66411
![]() |
Invitrogen™
C66411 |
Contains enough electrocompetent cells for 25 x 40μL rxns or 50 x 20μL rxns | 10 x 100 μL |
Each for $530.00
|
|
Description
- Allows stable replication of high-copy number plasmids
- hsdR for efficient transformation of unmethylated DNA from PCR amplifications
- mcrA for efficient transformation of methylated DNA from genomic preparations
- lacZΔM15 for blue/white color screening of recombinant clones
- endA1 for cleaner preparations of DNA and better results in downstream applications due to elimination of non-specific digestion by Endonuclease I
- recA1 for reduced occurrence of non-specific recombination in cloned DNA
- Bacterial/Yeast Strain: TOP10
Applications:
Cloning, Transformation
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.