Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOP1MT Mouse anti-Human, Clone: TOP1MT/488, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14239

Catalog No. 89-034-323


Add to cart

Description

Description

Mouse monoclonal antibody raised against full length recombinant human TOP1MT.

  • High specificity
  • High reactivity
Specifications

Specifications

TOP1MT
TOP1MT/488
Mouse monoclonal antibody raised against full length recombinant human TOP1MT.
In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Liquid
TOP1MT
Recombinant protein corresponding to full length human TOP1MT.
Protein A/G purification
RUO
Primary
116447
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Q969P6
2900052H09Rik
Mouse
IgG2b, kappa
100 ug
Store at 4°C.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)