Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOP1MT Rabbit anti-Human, Clone: TOP1MT/2883R, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30756810UG

 View more versions of this product

Catalog No. NB122609


Add to cart

Description

Description

TOP1MT Monoclonal specifically detects TOP1MT in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

TOP1MT
TOP1MT/2883R
Unconjugated
10 mM PBS with 0.05% BSA
mitochondrial, mitochondrial DNA topoisomerase I, topoisomerase (DNA) I, mitochondrial
Recombinant full-length human TOP1MT protein
Protein A or G purified
RUO
Primary
116447
Immunohistochemistry (Paraffin)
0.2 mg/ml
Immunohistochemistry-Paraffin 1-2 ug/ml
Purified
Rabbit
IgG
100 μg
Store at 4°C.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit