Learn More
TOP2A Mouse anti-Human, Clone: OTI2A9, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF802384
Catalog No. 50-166-5975
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Topo II alpha plays important roles in synthesis and transcription of DNA as well as chromosomal segregation during mitosis. It is reported to be a sensitive and specific marker of late S-, G2- & M-phases in transformed and developmentally regulated normal cellsTopo II alpha is also implicated in drug resistance of tumor cells.
Specifications
Specifications
TOP2A
Monoclonal
Unconjugated
TOP2A
Lyophilized
TOP2A
Human recombit protein fragment corresponding to amino acids 1100-1531 of human TOP2A produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI2A9
PBS with 8% trehalose and no preservative; pH 7.3
P11388
TOP2, TP2A
Mouse
IgG2b
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
7153